missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRAM Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00 € - 462.00 €
Specifications
| Antigen | DRAM |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
DRAM Polyclonal antibody specifically detects DRAM in Mouse samples. It is validated for Western BlotSpecifications
| DRAM | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS with 50% glycerol, pH7.3. | |
| 55332 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| Damage-regulated autophagy modulator, DNA damage-regulated autophagy modulator protein 1, DNA-damage regulated autophagy modulator 1, DRAMFLJ11259 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human DRAM1 (NP_060840.2). GLVGCFGMGIVANFQELAVPVVHDGGALLAFVCGVVYTLLQSIISYKSCPQWNSLSTCHIRMVISAVSCAAVIPMIVCASLISITKLEWNPREKDYVYHVV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title