missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRAM Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92236-0.1ml
This item is not returnable.
View return policy
Description
DRAM Polyclonal antibody specifically detects DRAM in Mouse samples. It is validated for Western Blot
Specifications
| DRAM | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| Damage-regulated autophagy modulator, DNA damage-regulated autophagy modulator protein 1, DNA-damage regulated autophagy modulator 1, DRAMFLJ11259 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human DRAM1 (NP_060840.2). GLVGCFGMGIVANFQELAVPVVHDGGALLAFVCGVVYTLLQSIISYKSCPQWNSLSTCHIRMVISAVSCAAVIPMIVCASLISITKLEWNPREKDYVYHVV | |
| 0.1 mL | |
| Cancer | |
| 55332 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction