missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DRAQ5™ Fluorescent Probe Solution (5 mM)

Product Code. 11863865 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
200 μL
1 mL
Unit Size:
1mL
200µL
50µL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11863865 200 μL 200µL
11895005 1 mL 1mL
11820154 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11863865 Supplier Thermo Scientific™ Supplier No. 62251
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stain live or fixed cells with this far-red DNA stain.

Stain live or fixed cells with Thermo Scientific™ DRAQ5 Fluorescent Probe, a far-red DNA stain.

Fluorescence cell-based assays, such as flow cytometry, in-cell ELISA, fluorescence microscopy and high-content imaging require a fluorescent label to identify individual cells. When using multiple fluorescent probes to detect different cellular targets or activities, each probe must have a fluorescent spectrum different than the other probes. The blue-fluorescent DNA-binding probes, Hoechst and DAPI are frequently used; however, these probes cannot be used when UV illumination is unavailable or other blue-emitting fluorescent probes are used. Therefore, nuclear probes that emit in a color other than blue is useful for cell identification and counting, and for determining nuclear morphology and DNA content.
  • Due to its far-red excitation and emission, the DRAQ5 Fluorescent Probe can be multiplexed with many other fluorophores and is ideal for cells expressing green fluorescent protein (GFP) fusion proteins
  • Compatible with many existing protocols across a wide range of instrumentation platforms
  • Emits in the far-red region, is lipophilic and crosses cell and nuclear membranes in live and fixed cells and tissues for rapid DNA staining
  • Stain is water-soluble, supplied ready-to-use and does not require RNase, cell lysis, or a washing step, making it compatible with automation
  • Because DNA staining is stoichiometric, the DRAQ5 Fluorescent Probe can be used for DNA content analysis in cell proliferation studies
TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Cell-permeant
Color Red
Content And Storage Store at 4°C.
Description DRAQ5 Fluorescent Probe Solution (5 mM)
Detection Method Fluorescence
For Use With (Application) Cell Imaging
For Use With (Equipment) Fluorescence Microscope, High Content Analysis Instrument
Product Type Fluorescent Probe Solution
Dye Type Cell-Permeant
Form Liquid
Product Line Pierce
Purity or Quality Grade > 99%
Quantity 200 μL
Solubility Soluble at working concentrations in water and typical buffers
Sub Cellular Localization Nucleus, Cytosol, Cytoplasm
Excitation Wavelength 647 nm
Emission Wavelength 681
Show More Show Less
compliance-icons
Product Identifier
  • DRAQ5 Fluorescent Probe Solution (5 mM)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 1,5-bis{[2-(di-methylamino) ethyl]amino}-4, 8-dihydroxyanthracene-9,10-dione

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.