missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRB1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92131-0.1ml
This item is not returnable.
View return policy
Description
DRB1 Polyclonal antibody specifically detects DRB1 in Mouse samples. It is validated for Western Blot
Specifications
| DRB1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| developmentally regulated RNA-binding protein 1, Developmentally-regulated RNA-binding protein 1, DRB1FLJ44612, DRBP1, MGC42237, putative RNA binding protein RB-1, RB-1, RNA binding motif protein 45, RNA-binding motif protein 45, RNA-binding protein 45 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human RBM45 (NP_694453.2). MDEAGSSASGGGFRPGVDSLDEPPNSRIFLVISKYTPESVLRERFSPFGDIQDIWVVRDKHTKESKGIAFVKFARSSQACRAMEEMHGQCLGPNDTKPIKVFIAQSRSSGSHRDVEDEELTRIFVMIPKSYTEEDLREKFKVYGDIEYCSIIKNKVTGESKGLGYVRYLKPSQAAQAIENCDRSFRAILAEPKNKASESSEQDYYSNMRQEALGHEPRVNMFPFEQQSEFSSFDKNDSRGQEAISKRLSVVSRVPFTEEQLFSIFDIVPGLEYCEVQRDPYSNYGHGVVQYFNVASAIYA | |
| 0.1 mL | |
| Cell Biology | |
| 129831 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur