missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRB1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00 € - 462.00 €
Specifications
| Antigen | DRB1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
DRB1 Polyclonal antibody specifically detects DRB1 in Mouse samples. It is validated for Western BlotSpecifications
| DRB1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS with 50% glycerol, pH7.3. | |
| 129831 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| developmentally regulated RNA-binding protein 1, Developmentally-regulated RNA-binding protein 1, DRB1FLJ44612, DRBP1, MGC42237, putative RNA binding protein RB-1, RB-1, RNA binding motif protein 45, RNA-binding motif protein 45, RNA-binding protein 45 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human RBM45 (NP_694453.2). MDEAGSSASGGGFRPGVDSLDEPPNSRIFLVISKYTPESVLRERFSPFGDIQDIWVVRDKHTKESKGIAFVKFARSSQACRAMEEMHGQCLGPNDTKPIKVFIAQSRSSGSHRDVEDEELTRIFVMIPKSYTEEDLREKFKVYGDIEYCSIIKNKVTGESKGLGYVRYLKPSQAAQAIENCDRSFRAILAEPKNKASESSEQDYYSNMRQEALGHEPRVNMFPFEQQSEFSSFDKNDSRGQEAISKRLSVVSRVPFTEEQLFSIFDIVPGLEYCEVQRDPYSNYGHGVVQYFNVASAIYA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title