missing translation for 'onlineSavingsMsg'
Learn More

DRO Mixture 2000 μg/mL in Dichloromethane/Hexane, Dr. Ehrenstorfer

Product Code. 30310479
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30310479 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310479 Supplier Dr. Ehrenstorfer Supplier No. DREA50000286DX
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material n-Decane∣n-Docosane∣n-Dodecane∣n-Eicosane∣n-Hexacosane∣n-Hexadecane∣n-Octadecane∣n-Tetradecane∣Octacosane∣Tetracosane
CAS 124-18-5, 629-97-0, 112-40-3, 112-95-8, 630-01-3, 544-76-3
Analyte or Component Names n-Decane∣n-Docosane∣n-Dodecane∣n-Eicosane∣n-Hexacosane∣n-Hexadecane∣n-Octadecane∣n-Tetradecane∣Octacosane∣Tetracosane
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane/Hexane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) n-Decane 2000 μg/mL∣n-Docosane 2000 μg/mL∣n-Dodecane 2000 μg/mL∣n-Eicosane 2000 μg/mL∣n-Hexacosane 2000 μg/mL∣n-Hexadecane 2000 μg/mL∣n-Octadecane 2000 μg/mL∣n-Tetradecane 2000 μg/mL∣Octacosane 2000 μg/mL∣Tetracosane 2000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.