Learn More
Thermo Scientific™ dU Phosphoramidite, standard grade, serum vial bottle
dU CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyuridine (dU) for use in oligonucleotide synthesis.
12.30 € - 106.00 €
Specifications
For Use With (Equipment) | AKTA Synthesizers, Applied Biosystems Synthesizers |
---|---|
Product Type | dU CEP |
Content And Storage | Store in freezer (at or below -20°C). |
Nucleoside Type | Deoxyuridine (dU) |
Shipping Condition | Room Temperature |
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
17977132
|
Thermo Scientific™
27-1738-02 |
106.00 €
1g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18044090
|
Thermo Scientific™
27-1738-03 |
12.30 €
|
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
dU CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyuridine (dU) for use in oligonucleotide synthesis. The incorporation of a deoxyuridine base into a DNA sequence can be utilized to induce mutagenic effects. Enzymatically the uracil is removed, creating an abasic site at the dU position, and thus generating site-specific breaks in the oligo. This product is supplied in a 20 mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers of oligonucleotide synthesizers.
Features:
• Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C39H47N4O8P
• Base: dU
• Formula weight: 730.8 g/mol
• CAS number: 109389-30-2
• HPLC Purity: ≥98.0%; conforms to reference
• 31P NMR Purity: ≥98.0%; conforms to reference
Specifications
AKTA Synthesizers, Applied Biosystems Synthesizers | |
Store in freezer (at or below -20°C). | |
Room Temperature | |
Phosphoramidite | |
DNA |
dU CEP | |
Deoxyuridine (dU) | |
98% | |
Standard | |
Powder |
For Research Use Only. Not for use in diagnostic procedures.