missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Dulbecco Medium
BIOPHARMA

Product Code. 15353781
Change view
Click to view available options
Quantity:
10 L
Formulation:
Amino acids, vitamins, 4.5 g/L glucose
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Formulation Unit Size
15353781 10 L Amino acids, vitamins, 4.5 g/L glucose 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15353781 Supplier Corning™ Supplier No. 90013PB
  • ONLINE EXCLUSIVE PRICE
    56.05 €
    49.23 € / 10 L
    Save 6.82 €
    12% Off
 Restricted Item
Estimated Shipment: 01-04-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To assess growth promotion and cytotoxicity characteristics, Corning Classical Media products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis. Product performance is measured by harvest-to-plant ratios or fold increase according to predetermined specifications. Liquid and powder classical media products are also tested to determine pH and osmolality. Classical media powders are also tested for residual moisture. The pH and osmolality tests are performed according to specific procedures in which all equipment is calibrated using standards traceable to the National Institute of Standards and Technology. Biological testing of Corning liquid classical media products guarantees low levels of endotoxin, mycoplasma, and an absence of contamination. Endotoxins: classical media products are tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) Chromogenic Assay. For release, each lot of standard liquid classical media must have an endotoxin level of less than 0.25 EU/mL. Classical media powder products must have an endotoxin level of less than 0.25 EU/mL when reconstituted to formula weight with low endotoxin water. All Corning liquid classical media products are tested for sterility and liquid classical media products are tested for mycoplasma off-site using a large-volume method.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Primary cultures of mouse and chicken cells
Product Type Dulbecco Medium
Sterility Sterile
For Use With (Application) Primary Cell Culture
Quantity 10 L
Certifications/Compliance Yes
Endotoxin Level < 0.25 EU/mL
Formulation Amino acids, vitamins, 4.5 g/L glucose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.