missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Dulbecco's Phosphate Buffered Saline
BIOPHARMA

Product Code. 15343571
Change view
Click to view available options
Quantity:
10,000 mL
20,000 mL
6 x 500 mL
Concentration:
10X
1X
Unit Size:
10 L
20 L
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Concentration unitSize
15343571 6 x 500 mL 10X Pack of 6
15353631 20,000 mL 1X 20 L
15363631 10,000 mL 1X 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15343571 Supplier Corning™ Supplier No. 20031CV
  • ONLINE EXCLUSIVE PRICE
    121.05 €
    107.82 € / Pack of 6
    Save 13.23 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

For use alone or combined with other agents for washing tissues and cells

  • Provide cells with water and inorganic ions
  • Maintains physiological pH and osmotic pressure
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DPBS
Concentration 10X
Quantity 6 x 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.