missing translation for 'onlineSavingsMsg'
Learn More

Duloxetine hydrochloride, MedChemExpress

Product Code. 16488136
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488136 100 mg 100mg
16484257 50 mg 50mg
16468368 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488136 Supplier Medchem Express Supplier No. HYB0161A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Duloxetine hydrochloride ((S)-Duloxetine hydrochloride) is a serotonin-norepinephrine reuptake inhibitor (SNRI) with a Ki of 4.6 nM, used for treatment of major depressive disorder and generalized anxiety disorder (GAD).

Duloxetine hydrochloride ((S)-Duloxetine hydrochloride) is a serotonin-norepinephrine reuptake inhibitor (SNRI) with a Ki of 4.6 nM, used for treatment of major depressive disorder and generalized anxiety disorder (GAD).

Specifications

Chemical Name or Material Duloxetine hydrochloride
CAS 136434-34-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H20ClNOS
Quantity 100 mg
Synonym (S)-Duloxetine hydrochloride LY-248686 hydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (299.51 mM) ∣H2O : 5 mg/mL (14.98 mM; Need ultrasonic)
SMILES CNCC[C@H](OC1=CC=CC2=C1C=CC=C2)C3=CC=CS3.[H]Cl
Molecular Weight (g/mol) 333.88
Formula Weight 333.88
Percent Purity 99.09%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.