Learn More
DuraCal™ 4.01 Value pH Buffer, Hamilton™
Product Code. 10714673
Change view
Click to view available options
Quantity:
3 x 500 mL
Unit Size:
Pack of 3
Product Code. Quantity unitSize
10714673 3 x 500 mL Pack of 3
1 options
Product Code. 10714673 Supplier Hamilton Supplier No. 238917
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Economical choice since only about 20mL of buffer is used per calibration. Hamilton™ DuraCal™ 4.01 Value pH Buffers have actual values determined by a DKD laboratory, accredited for pH measurement.
  • 4.01 pH value
  • Immune to micro-organisms
  • Expiration date on the bottle
  • First class certificate with traceability to international standards
Specifications
pH 4.01
Quantity 3 x 500 mL