missing translation for 'onlineSavingsMsg'
Learn More

DuraCal™ pH Buffer Set, Hamilton™

Product Code. 10081374
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
1500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10081374 500 mL 1500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10081374 Supplier Hamilton Supplier No. 238924
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Economical choice since only about 20mL of buffer is used per calibration. Hamilton™ DuraCal™ pH Buffer Sets have actual values determined by a DKD laboratory, accredited for pH measurement.

  • Includes one 500mL bottle of each value: 4.01, 7.00 and 10.01
  • Immune to micro-organisms
  • Expiration date on the bottle
  • First class certificate with traceability to international standards

Specifications

pH 4.01, 7.00, 10.01
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.