missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DUSP13 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA141642

Product Code. 11534630

  • 520.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Suggested positive control: human skeletal muscle lysate. 93% sequence identity with Mouse.

DUSP13, a novel DSP, is a member of highly conserved Protein-tyrosine phosphatase super family. Members of this family cooperate with protein kinases and play a vital role in cell cycle regulation. DUSP13 consists of a conserved catalytic domain and is expressed specifically in the testis and skeletal muscle. Based on mRNA localization and expression studies during development, DUSP13 is suggested to be expressed as 3 isoforms due to alternative splicing. Reports suggest DUSP13 might be involved in the regulation of meiosis and differentiation of testicular germ cells during spermatogenesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DUSP13
Polyclonal
Unconjugated
DUSP13
BEDP; Branching-enzyme interacting DSP; branching-enzyme interacting dual-specificity protein phosphatase; Dsp13-like protein; dual specificity phosphatase 13; Dual specificity phosphatase SKRP4; dual specificity protein phosphatase 13; Dual specificity protein phosphatase 13 isoform A; Dual specificity protein phosphatase 13 isoform B; dual specificity protein phosphatase isoform MDSP; dual specificity tyrosine phosphatase TS-DSP6; dual-specificity tyrosine phosphatase TS-DSP6; DUSP13; Dusp13a; DUSP13B; FLJ32450; Gm1203; LMW-DSP6; Mdsp; muscle-restricted DSP; muscle-restricted dual specificity phosphatase; muscle-restricted dual specificity phosphatase; testis and skeletal muscle-specific dual specificity phosphatase; RP11-487I5.6; SKRP4; testis- and skeletal muscle-specific DSP; testis and skeletal muscle-specific dual specificity phosphatase; Testis- and skeletal-muscle-specific DSP; TMDP; TS-DSP6
Rabbit
Protein G
RUO
361002, 489054, 51207, 616048
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
Q6B8I1, Q9UII6
DUSP13, DUSP13B
A portion of amino acids 150-200 of human DUSP13.
100 μg
Primary
Bovine, Canine, Human, Monkey, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.