missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUSP13 Polyclonal Antibody

Description
Immunogen sequence: MDSLQKQDLR RPKIHGAVQA SPYQPPTLAS LQRLLWVRQA ATLNHIDEVW PSLFLGDAYA ARDKSKLIQL GITHVVNAAA GKFQVDTGAK FYRGMSLEYY GIEADDNPFF DLSVYFLPVA RYIRAALSVP QGRVLVHCAM GVSRSATLVL AFLMICENMT LVEAIQTVQA HRNICPNSGF LRQLQVLDNR LGRETGRF; Positive Samples: Mouse skeletal muscle, Rat skeletal muscle; Cellular Location: Cytoplasm.
DUSP13, a novel DSP, is a member of highly conserved Protein-tyrosine phosphatase super family. Members of this family cooperate with protein kinases and play a vital role in cell cycle regulation. DUSP13 consists of a conserved catalytic domain and is expressed specifically in the testis and skeletal muscle. Based on mRNA localization and expression studies during development, DUSP13 is suggested to be expressed as 3 isoforms due to alternative splicing. Reports suggest DUSP13 might be involved in the regulation of meiosis and differentiation of testicular germ cells during spermatogenesis.
Specifications
Specifications
| Antigen | DUSP13 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.19 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | DUSP13 |
| Gene Accession No. | Q6B8I0 |
| Gene Alias | BEDP; Branching-enzyme interacting DSP; branching-enzyme interacting dual-specificity protein phosphatase; Dsp13-like protein; dual specificity phosphatase 13; Dual specificity phosphatase SKRP4; dual specificity protein phosphatase 13; Dual specificity protein phosphatase 13 isoform A; Dual specificity protein phosphatase 13 isoform B; dual specificity protein phosphatase isoform MDSP; dual specificity tyrosine phosphatase TS-DSP6; dual-specificity tyrosine phosphatase TS-DSP6; DUSP13; Dusp13a; DUSP13B; FLJ32450; Gm1203; LMW-DSP6; Mdsp; muscle-restricted DSP; muscle-restricted dual specificity phosphatase; muscle-restricted dual specificity phosphatase; testis and skeletal muscle-specific dual specificity phosphatase; RP11-487I5.6; SKRP4; testis- and skeletal muscle-specific DSP; testis and skeletal muscle-specific dual specificity phosphatase; Testis- and skeletal-muscle-specific DSP; TMDP; TS-DSP6 |
| Gene Symbols | DUSP13 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?