missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DUSP19 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17227913 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17227913 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17227913 Supplier Invitrogen™ Supplier No. PA5115147

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DUSP19.

Dual-specificity phosphatases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP19
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene DUSP19
Gene Accession No. Q8WTR2
Gene Alias 5930436K22Rik; C79103; dual specificity phosphatase 19; dual specificity phosphatase TS-DSP1; Dual specificity protein phosphatase 19; DUSP17; DUSP19; LMWDSP3; LMW-DSP3; low molecular weight dual specificity phosphatase 3; Protein phosphatase SKRP1; SAPK pathway-regulating phosphatase 1; SKRP1; Stress-activated protein kinase pathway-regulating phosphatase 1; TS-DSP1
Gene Symbols DUSP19
Host Species Rabbit
Immunogen A synthesized peptide derived from human DUSP19(Accession Q8WTR2), corresponding to amino acid residues K113-G163.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 142679
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.