missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 539.00 €
Specifications
| Antigen | DUSP8 |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18421931
|
Novus Biologicals
NBP1-88385-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18281776
|
Novus Biologicals
NBP1-88385 |
0.1 mL |
539.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DUSP8 Polyclonal specifically detects DUSP8 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| DUSP8 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| C11orf81, chromosome 11 open reading frame 81, dual specificity phosphatase 8, Dual specificity protein phosphatase hVH-5, EC 3.1.3.16, EC 3.1.3.48, FLJ42476, FLJ42958, HVH8, serine/threonine specific protein phosphatase, vaccinia virus homolog, VH5 | |
| DUSP8 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Hypoxia, Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 1850 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SDDAYRFVKDRRPSISPNFNFLGQLLEYERSLKLLAALQGDPGTPSGTPEPPPSPAAGAPLPRLPPPTSESAATGNAAAR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title