missing translation for 'onlineSavingsMsg'
Learn More

Dutasteride, MedChemExpress

Product Code. 16481927
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481927 50 mg 50mg
16476246 100 mg 100mg
16405778 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481927 Supplier Medchem Express Supplier No. HY13613
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dutasteride (GG745) is a potent inhibitor of both 5α-reductase isozymes. Dutasteride may possess off-target effects on the androgen receptor (AR) due to its structural similarity to DHT.

Dutasteride (GG745) is a potent inhibitor of both 5α-reductase isozymes. Dutasteride may possess off-target effects on the androgen receptor (AR) due to its structural similarity to DHT.

Specifications

Chemical Name or Material Dutasteride
Physical Form Solid
CAS 164656-23-9
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C27H30F6N2O2
For Use With (Application) Cancer-programmed cell death
Synonym GG 745 GI 198745
Solubility Information DMSO : 33.33 mg/mL (63.06 mM; Need ultrasonic)
SMILES O=C([C@H]1CC[C@]2([H])[C@]1(C)CC[C@]3([H])[C@@]4(C)C=CC(N[C@]4([H])CC[C@]32[H])=O)NC5=CC(C(F)(F)F)=CC=C5C(F)(F)F
Molecular Weight (g/mol) 528.53
Formula Weight 528.53
Percent Purity 99.73%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.