missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dUTPase Antibody (SR1280), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22634-100ul
This item is not returnable.
View return policy
Description
dUTPase Monoclonal antibody specifically detects dUTPase in Human samples. It is validated for Western Blot, Immunofluorescence, Serum
Specifications
| dUTPase | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR1280 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence, Serum 1:50 | |
| deoxyuridine triphosphatase, dUTP pyrophosphatasedUTP nucleotidohydrolase, dUTPasedeoxyuridine 5'-triphosphate nucleotidohydrolase, mitochondrial, EC 3.6.1.23, FLJ20622 | |
| A synthesized peptide derived from human dUTPase (Uniprot #: P33316) | |
| 100 μg | |
| Lipid and Metabolism | |
| 1854 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction