missing translation for 'onlineSavingsMsg'
Learn More

Dyclonine hydrochloride, MedChemExpress

Product Code. p-200095907
Encompass_Preferred
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 16438458

Brand: Medchem Express HYB0364A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dyclonine hydrochloride (Dyclocaine hydrochloride) is an effective component of Runhou tablets. Dyclonine hydrochloride has significant bactericidal and fungicidal activity.

Dyclonine hydrochloride (Dyclocaine hydrochloride) is an effective component of Runhou tablets. Dyclonine hydrochloride has significant bactericidal and fungicidal activity.

Specifications

Chemical Name or Material Dyclonine hydrochloride
CAS 536-43-6
Color White
Purity Grade Notes Research
Molecular Formula C18H28ClNO2
Quantity 10 g
Synonym Dyclocaine hydrochloride
Solubility Information H2O : 50 mg/mL (153.44 mM; Need ultrasonic) ∣DMSO : ≥ 25 mg/mL (76.72 mM)
SMILES O=C(C1=CC=C(OCCCC)C=C1)CCN2CCCCC2.Cl
Molecular Weight (g/mol) 325.87
Formula Weight 325.87
Percent Purity 98.15%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.