missing translation for 'onlineSavingsMsg'
Learn More

DYNC1H1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

292.00 € - 572.00 €

Specifications

Antigen DYNC1H1
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18486600
View Documents
Novus Biologicals
NBP1-84254-25ul
25 μL
292.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18268066
View Documents
Novus Biologicals
NBP1-84254
0.1 mL
572.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DYNC1H1 Polyclonal specifically detects DYNC1H1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

DYNC1H1
Polyclonal
Rabbit
Cell Cycle and Replication
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
1778
This antibody was developed against Recombinant Protein corresponding to amino acids:RSLETCMYDHKTFSEILNRVQKAVDDLNLHSYSNLPIWVNKLDMEIERILGVRLQAGLRAWTQVLLGQAEDKAEVDMDTDAPQVSHKPGGEPKIKNVVHELRITNQVIYLNPPIEECRYKLYQEMFAWKMVVLSLPRIQSQR
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
cytoplasmic dynein 1 heavy chain 1, Cytoplasmic dynein heavy chain 1, DHC1a, DHC1Dynein heavy chain, cytosolic, DKFZp686P2245, Dnchc1, DNCLDNCH1, DNECLDYHC, dynein, cytoplasmic 1, heavy chain 1, dynein, cytoplasmic, heavy polypeptide 1, HL-3, KIAA0325, p22
DYNC1H1
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.