missing translation for 'onlineSavingsMsg'
Learn More

DZ/T 0064.72-2021 Organophosphorus Pesticide Mixture 696 1000 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30318069
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30318069 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318069 Supplier Dr. Ehrenstorfer Supplier No. DREA50000696AC

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorpyrifos∣Dichlorvos∣Dimethoate∣Malathion∣Parathion-ethyl∣Parathion-methyl∣Phorate
CAS 2921-88-2, 62-73-7, 60-51-5, 121-75-5, 56-38-2, 298-00-0
Analyte or Component Names Chlorpyrifos∣Dichlorvos∣Dimethoate∣Malathion∣Parathion-ethyl∣Parathion-methyl∣Phorate
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetone
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Chlorpyrifos 1000 μg/mL∣Dichlorvos 1000 μg/mL∣Dimethoate 1000 μg/mL∣Malathion 1000 μg/mL∣Parathion-ethyl 1000 μg/mL∣Parathion-methyl 1000 μg/mL∣Phorate 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.