missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DZIP1 Antibody (05), Novus Biologicals™
Mouse Monoclonal Antibody
220.00 € - 326.00 €
Specifications
| Antigen | DZIP1 |
|---|---|
| Clone | 05 |
| Dilution | Immunohistochemistry-Paraffin 1:100-1:500 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18677368
|
Novus Biologicals
NBP3-05837-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640348
|
Novus Biologicals
NBP3-05837-50ul |
50 μg |
220.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DZIP1 Monoclonal antibody specifically detects DZIP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| DZIP1 | |
| Immunohistochemistry-Paraffin 1:100-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 22873 | |
| IgG2a | |
| Protein A purified |
| 05 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| DAZ interacting protein 1, DAZ-interacting protein 1/2, DZIP, DZIP2, DZIPDAZ interacting protein testis1, DZIPt1, KIAA0996zinc finger DAZ interacting protein 1, RP11-23E3.3, zinc finger protein DZIP1, zinc-finger protein DZIPt1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with a synthetic peptide corresponding to the center region of the Human DZIP1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title