missing translation for 'onlineSavingsMsg'
Learn More

(E)-3-(3-(Trifluoromethyl)phenyl)acrylaldehyde (>90%), TRC

Product Code. 30456727
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456727 5 g 5g
30451457 1 g 1g
30453731 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456727 Supplier Toronto Research Chemicals Supplier No. TRCT7916205G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 262268-58-6
Molecular Formula C10H7F3O
Molecular Weight (g/mol) 200.16
Synonym (2E)-3-[3-(Trifluoromethyl)phenyl]-2-propenal, (2E)-3-[3-(Trifluoromethyl)phenyl]prop-2-enal
IUPAC Name (E)-3-[3-(trifluoromethyl)phenyl]prop-2-enal
SMILES FC(F)(F)c1cccc(\C=C\C=O)c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H7F3O/c11-10(12,13)9-5-1-3-8(7-9)4-2-6-14/h1-7H/b4-2+
Formula Weight 200.0449
Chemical Name or Material (E)-3-(3-(Trifluoromethyl)phenyl)acrylaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.