missing translation for 'onlineSavingsMsg'
Learn More

(E)-3,4-Dihydroxybenzylideneacetone, 97%

Product Code. 15443327
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15443327 1 g 1g
15453327 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15443327 Supplier Thermo Scientific Alfa Aesar Supplier No. H25901.03
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 123694-03-1 | C10H10O3 | 178.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123694-03-1
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD00916691
InChI Key YIFZKRGUGKLILR-UHFFFAOYSA-N
Synonym 3-buten-2-one,4-3,4-dihydroxyphenyl-, 3e, 3,4-dihydroxybenzalacetone, acmc-20dpit
PubChem CID 53446072
IUPAC Name 4-(3,4-dihydroxyphenyl)but-3-en-2-one
SMILES CC(=O)C=CC1=CC(=C(C=C1)O)O

Specifications

Melting Point 173°C to 175°C
Quantity 1 g
Formula Weight 178.19
Percent Purity 97%
Chemical Name or Material (E)-3,4-Dihydroxybenzylideneacetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.