missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(E)-9-Hexadecenoic acid, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Palmitelaidic Acid |
| CAS | 10030-73-6 |
| Analyte or Component Names | Palmitelaidic Acid |
| Shipping Condition | +20°C |
| Molecular Formula | C16 H30 O2 |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | 9-Hexadecenoic acid, (E)- (8CI), (9E)-9-Hexadecenoic acid, (E)-9-Hexadecenoic acid, (E)-Hexadec-9-enoic acid, 9-trans-Hexadecenoic acid, Palmitelaidic acid, trans-9-Hexadecenoic acid, trans-Palmitoleic acid, 9-Hexadecenoic acid, (9E)-, (E)-9-Hexadecenoic Acid |
| InChI Formula | InChI=1S/C16H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h7-8H,2-6,9-15H2,1H3,(H,17,18)/b8-7+ |
| SMILES | CCCCCC\C=C\CCCCCCCC(=O)O |
| IUPAC Name | (E)-hexadec-9-enoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?