missing translation for 'onlineSavingsMsg'
Learn More

(E)-alpha-Methylstilbene, 98%

Product Code. 11448276
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448276 5 g 5g
11458276 25 g 25g
11468276 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448276 Supplier Thermo Scientific Alfa Aesar Supplier No. A15351.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 833-81-8 | C15H14 | 194.277 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 833-81-8
Molecular Formula C15H14
Molecular Weight (g/mol) 194.277
MDL Number MFCD00026343
InChI Key OVZXISBUYCEVEV-OUKQBFOZSA-N
Synonym 1,2-diphenylpropene, e-alpha-methylstilbene, .alpha.-methylstilbene, 1,2-diphenyl-1-propene, 1-methyl-1,2-diphenylethene, stilbene, .alpha.-methyl, 1-propene, 1,2-diphenyl, 1-methyl-2-phenylvinyl benzene, trans-alpha-methylstilbene, chembl14773
PubChem CID 1549166
IUPAC Name [(E)-1-phenylprop-1-en-2-yl]benzene
SMILES CC(=CC1=CC=CC=C1)C2=CC=CC=C2

Specifications

Melting Point 79°C to 82°C
Density 0.986
Quantity 5 g
Beilstein 1906425
Formula Weight 194.28
Percent Purity 98%
Chemical Name or Material (E)-alpha-Methylstilbene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.