missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Cadherin Antibody (180224), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB18381H
This item is not returnable.
View return policy
Description
E-Cadherin Monoclonal antibody specifically detects E-Cadherin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
Specifications
| E-Cadherin | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
| 180224 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready | |
| Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin | |
| Mouse myeloma cell line NS0-derived recombinant human E-Cadherin, Asp155-Ile707, Accession # P12830 | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction | |
| 999 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction