missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-cadherin Monoclonal Antibody (ECCD-2)
GREENER_CHOICE

Product Code. 10199532
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
Product Code. Quantity unitSize
10199532 100 μg Each
1 options
This item is not returnable. View return policy

Product Code. 10199532

Brand: Invitrogen™ 131900

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Rat Monoclonal Antibody

Source: Monoclonal antibody was obtained by fusing the P3-X63-Ag8 mouse myeloma cell line with spleen cells of Donryu rat after immunization with mouse teratocarcinoma and mouse liver E-cadherin fragment. Specificity: This antibody specifically reacts with mouse E-cadherin. This antibody does not inhibit E-cadherin-dependent cell-cell contact. Cross reactivity: This antibody reacts with human and dog E-cadherin. This antibody does not react with bovine or chicken E-cadherin. Western blot can be performed under reducing or non-reducing conditions. Reconstitution and dilution: dissolve the lyophilized antibody in 50 μl of distilled water to a final concentration 2.0 mg/mL. This solution can be used as a stock solution. If dilution is necessary for your application, dilute the stock solution in 500 μL or more of the following dilution solution: 20mM TBS (pH 7.5), 10mM CaCl2, 1.0% BSA, 0.1% NaN3. Note that TBS should be used for dilution, as PBS causes precipitation. Storage: When stored at 4°C, 0.1% sodium azide should be added as a preservative.Store the antibody at a minimum concentration of 2.0 mg/mL; a lower antibody concentration may result in decreased stability.The stock solution (2.0mg/mL) should be stored in aliquots at -20°C for 1year, or stored at 4°C for 6 months after adding 0.1% sodium azide. Avoid repeated freeze-thaw cycles. Diluted antibody should not be stored.

E-Cadherin (epithelial cadherin) is a classical cadherin from the cadherin (alcium dependent adhesion protein) superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid and ovarian cancer. Loss of function is thought to contribute to progression in cancer by increasing proliferation, invasion, and/or metastasis. The ectodomain of this protein mediates bacterial adhesion to mammalian cells and the cytoplasmic domain is required for internalization. Identified transcript variants arise from mutation at consensus splice sites. E-cadherin plays a central role in the growth and development of cells by controlling tissue architecture, and maintenance of tissue integrity. In humans, E-cadherin is encoded by the CDH1 gene present on chromosome 16. Studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E-cadherin
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone ECCD-2
Concentration 2.0 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA and no preservative; pH 7.5
Gene CDH1
Gene Accession No. P09803
Gene Alias AA960649; ARC-1; cadherin 1; cadherin 1, E-cadherin (epithelial); cadherin 1, epithelial; cadherin 1, type 1; cadherin 1, type 1, E-cadherin (epithelial); cadherin e; cadherin-1; cadherin-E; calcium-dependent adhesion protein, epithelial; CAM 120/80; CD324; CDH1; CDHE; cell adhesion molecule; cell-CAM 120/80; ECAD; E-cad; E-Cad/CTF1; E-Cad/CTF2; E-Cad/CTF3; Ecadherin; E-cadherin; E-cadherin 1; E-cadherin precursor; Epithelial cadherin; hab; half baked; LCAM; L-CAM; Um; UVO; Uvomorulin
Gene Symbols CDH1
Host Species Rat
Immunogen Mouse liver E-cadherin fragment.
Purification Method Ion-exchange chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12550
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.