missing translation for 'onlineSavingsMsg'
Learn More

(E,E)-5-[4-(Diethylamino)phenyl]penta-2,4-dienal, TRC

Product Code. 30385075
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385075 1 g -
30376049 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385075 Supplier Toronto Research Chemicals Supplier No. TRCD4436651G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868161-59-5
Molecular Formula C15H19NO
Molecular Weight (g/mol) 229.32
Synonym (2E,4E)-5-[4-(Diethylamino)phenyl]-2,4-pentadienal
SMILES CCN(CC)c1ccc(\C=C\C=C\C=O)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H19NO/c1-3-16(4-2)15-11-9-14(10-12-15)8-6-5-7-13-17/h5-13H,3-4H2,1-2H3/b7-5+,8-6+
Formula Weight 229.1467
Chemical Name or Material (E,E)-5-[4-(Diethylamino)phenyl]penta-2,4-dienal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.