missing translation for 'onlineSavingsMsg'
Learn More

(E,E)-Bisdemethoxycurcumin, TRC

Product Code. 30138271
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138271 250 mg 250mg
30003316 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138271 Supplier Toronto Research Chemicals Supplier No. TRCB425800250MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33171-05-0
Molecular Formula C19 H16 O4
Molecular Weight (g/mol) 308.33
Synonym 1,6-Heptadiene-3,5-dione, 1,7-bis(4-hydroxyphenyl)-, (1E,6E)-, 1,6-Heptadiene-3,5-dione, 1,7-bis(p-hydroxyphenyl)-, (E,E)- (8CI), (1E,6E)-1,7-Bis(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione, Bisdemethoxycurcumin, Bisdesmethoxycurcumin, Curcumin III, Didemethoxycurcumin
IUPAC Name (1E,6E)-1,7-bis(4-hydroxyphenyl)hepta-1,6-diene-3,5-dione
SMILES Oc1ccc(\C=C\C(=O)CC(=O)\C=C\c2ccc(O)cc2)cc1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C19H16O4/c20-16-7-1-14(2-8-16)5-11-18(22)13-19(23)12-6-15-3-9-17(21)10-4-15/h1-12,20-21H,13H2/b11-5+,12-6+
Formula Weight 308.1049
Percent Purity >95%
Chemical Name or Material Bisdemethoxycurcumin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.