missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Selectin/CD62E Antibody (RM0034-9J12), Alexa Fluor™ 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB110-85473AF488
This item is not returnable.
View return policy
Description
E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| E-Selectin/CD62E | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SELE | |
| Cytokine-stimulated mouse E-Selectin/CD62E | |
| 0.1 mL | |
| Immunology, Lipid and Metabolism | |
| 6401 | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0034-9J12 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, selectin E | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| This is specific for mouse E-Selectin | |
| Store at 4C in the dark. | |
| IgG2 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only