missing translation for 'onlineSavingsMsg'
Learn More

(E/Z)-FK-506 26,28-Allylic Ester Rearrangement Impurity (>90%), TRC

Product Code. 30008733
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008733 25 mg 25mg
30010827 100 mg 100mg
30008947 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008733 Supplier Toronto Research Chemicals Supplier No. TRCF37002025MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131944-48-4
Molecular Formula C44 H69 N O12
Molecular Weight (g/mol) 804.02
Synonym 17,21-Epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclopentacosine-1,9,22,23(6H,25H)-tetrone, 7,8,10,13,14,15,16,17,18,19,20,21,26,27,28,28a-hexadecahydro-7,21-dihydroxy-3-(4-hydroxy-3-methoxycyclohexyl)-16,18-dimethoxy-4,6,12,14,20-pentamethyl-10-(2-propenyl)-, [3R-[3R*(1R*,3R*,4R*),4E,6S*,7S*,10R*,11E,14S*,16S*,17R*,18S*,20R*,21R*,28aS*]]- (9CI), FK506-A, [3R-[3R*(1R*,3R*,4R*),4E,6S*,7S*,10R*,11E,14S*,16S*,17R*,18S*,20R*,21R*,28aS*]]-7,8,10,13,14,15,16,17,18,19,20,21,26,27,28,28a-Hexadecahydro-7,21-dihydroxy-3-(4-hydroxy-3-methoxycyclohexyl)-16,18-dimethoxy-4,6,12,14,20-pentamethyl-10-(2-propenyl)-17,21-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclopentacosine-1,9,22,23(6H,25H)-tetrone, Tacrolimus Impurity D, (E/Z)-FK-506 26,28-Allylic Ester Rearrangement Impurity
SMILES CO[C@@H]1C[C@@H](CC[C@H]1O)[C@H]2OC(=O)[C@@H]3CCCCN3C(=O)C(=O)[C@]4(O)O[C@H]([C@H](C[C@@H](C)C\C(=C\[C@@H](CC=C)C(=O)C[C@H](O)[C@@H](C)\C=C\2/C)\C)OC)[C@H](C[C@H]4C)OC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C44H69NO12/c1-10-13-30-19-25(2)18-26(3)20-37(54-8)40-38(55-9)22-29(6)44(52,57-40)41(49)42(50)45-17-12-11-14-32(45)43(51)56-39(31-15-16-33(46)36(23-31)53-7)28(5)21-27(4)34(47)24-35(30)48/h10,19,21,26-27,29-34,36-40,46-47,52H,1,11-18,20,22-24H2,2-9H3/b25-19+,28-21+/t26-,27-,29+,30+,31+,32-,33+,34-,36+,37-,38-,39-,40+,44+/m0/s1
Formula Weight 803.482
Chemical Name or Material Tacrolimus TLC Impurity 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.