missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ E2F6 Recombinant Protein Antigen

Product Code. 18120685 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 18120685

Brand: Novus Biologicals™ NBP269000PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified and high bioactivity. Generating reliable and reproducible results.

  • A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human E2F6
  • This peptide is useful as a blocking peptide for NBP2-69000.
  • Protein was purified by IMAC chromatography.
  • The expected concentration is greater than 0.5 mg/mL.
  • This product is intended to be used as a blocking antigen for antibody competition assays. Any other use of this antigen is done at the risk of the user. The use of this product for commercial production is strictly prohibited. Please contact technical support if you have any questions.
  • This product is for research use only and is not approved for use in humans or in clinical diagnosis.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 1876
Purification Method >80% by SDS-PAGE and Coomassie blue staining
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
For Use With (Application) Antibody Competition
Gene Symbol E2F6
Molecular Weight (g/mol) 26 kDa
Quantity 100 μL
Source E. coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-55058. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Storage Buffer PBS and 1M Urea, pH 7.4.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.