Learn More
Invitrogen™ E4BP4 (NFIL3) Monoclonal Antibody (MABA223), Alexa Fluor™ 647, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ 51-9812-41
Description
Description: This MABA223 monoclonal antibody recognizes human E4BP4 (NFIL3), a basic leucine zipper transcription factor first identified for its binding to the promoters of the adenovirus E4 protein and human IL-3. E4BP4 is involved in the transcriptional control of the mammalian circadian clock, motor neuron growth and survival, regulation of von Willbrand factor expression, and osteoblast function. In addition, it is required for the development of NK and NKT cells as well as CD8a+ dendritic cells. Although relatively high levels of mRNA are reported in resting NK and NKT cells, studies at eBioscience have observed expression of E4BP4 protein in lymphocytes only after activation. Applications Reported: This MABA223 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MABA223 antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells using the Foxp3/Transcription Factor Staining Buffer Set (Product No. 00-5523-00) and protocol. Please refer to Staining Intracellular Antigens for Flow Cytometry, Protocol B: One step protocol for intracellular (nuclear) proteins located at www.thermofisher.com/flowprotocols. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.
Expression of interleukin-3 (IL3; MIM 147740) is restricted to activated T cells, natural killer (NK) cells, and mast cell lines. Transcription initiation depends on the activating capacity of specific protein factors, such as NFIL3, that bind to regulatory regions of the gene, usually upstream of the transcription start site (Zhang et al., 1995 [PubMed 7565758]).
Specifications
| E4BP4 (NFIL3) | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Q16649 | |
| NFIL3 | |
| 21H2-hNFIL3 (aa 163-462). | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| MABA223 | |
| Alexa Fluor 647 | |
| NFIL3 | |
| AV225605; E4 promoter binding-protein 4; E4 promoter-binding protein 4; E4BP4; Embryo implantation-related NFIL3/E4BP4-like transcription factor; IL3BP1; Interleukin-3 promoter transcriptional activator; interleukin-3-binding protein 1; NF IL3A; NFIL3; NFIL3A; NF-IL3A; nuclear factor interleukin 3 regulated protein; nuclear factor interleukin-3-regulated protein; nuclear factor, interleukin 3 regulated; nuclear factor, interleukin 3, regulated; OTTHUMP00000021633; transcriptional activator NF-IL3A | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4783 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.