missing translation for 'onlineSavingsMsg'
Learn More

E7046, MedChemExpress

Product Code. 16410177
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16410177

Brand: Medchem Express HY103088

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress E7046 is an orally bioavailable and specific EP4 antagonist, with IC50 of 13.5 nM and Ki of 23.14 nM. E7046 exhibits anti-tumor activities.

E7046 is an orally bioavailable and specific EP4 antagonist, with IC50 of 13.5 nM and Ki of 23.14 nM. E7046 exhibits anti-tumor activities.

Specifications

Chemical Name or Material E7046
CAS 1369489-71-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H18F5N3O4
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (206.87 mM)
SMILES O=C(O)C1=CC=C([C@@H](NC(C2=C(OC3=CC=CC(C(F)(F)F)=C3)N(C)N=C2C(F)F)=O)C)C=C1
Molecular Weight (g/mol) 483.39
Formula Weight 483.39
Percent Purity 98.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.