missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E7046, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Description
E7046 is an orally bioavailable and specific EP4 antagonist, with IC50 of 13.5 nM and Ki of 23.14 nM. E7046 exhibits anti-tumor activities.
Specifications
Specifications
| Chemical Name or Material | E7046 |
| CAS | 1369489-71-3 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H18F5N3O4 |
| Quantity | 5 mg |
| Solubility Information | DMSO : ≥ 100 mg/mL (206.87 mM) |
| SMILES | O=C(O)C1=CC=C([C@@H](NC(C2=C(OC3=CC=CC(C(F)(F)F)=C3)N(C)N=C2C(F)F)=O)C)C=C1 |
| Molecular Weight (g/mol) | 483.39 |
| Formula Weight | 483.39 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction