missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EAAT2/GLT1 Antibody - Azide Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
Brand: Novus Biologicals NBP1-20136SS
This item is not returnable.
View return policy
Description
EAAT2/GLT1 Polyclonal specifically detects EAAT2/GLT1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, In-situ Hybridization, In vivo assay.
Specifications
| EAAT2/GLT1 | |
| Polyclonal | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:1000, In-situ Hybridization, In vivo assay | |
| P43006 | |
| SLC1A2 | |
| A synthetic peptide from mouse EAAT2/GLT1 conjugated to blue carrier protein was used as the antigen. The peptide is homologous in rat and human. | |
| 0.025 mL | |
| Hypoxia, Neuroscience | |
| 6506 | |
| Human, Mouse, Rat, Monkey | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Lyophilized from whole antisera with No Preservative | |
| GLT1, member 2, solute carrier family 1 (glial high affinity glutamate transporter), member 2, Solute carrier family 1 member 2 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction