Learn More
Thermo Scientific™ EASY-Spray™ HPLC Columns
Shop All Thermo Scientific ProductsDescription
Robust capillary flow HPLC columns for sensitive protein, peptide and mAbs separation using the stand alone or integrated easy to use design.
Ensure robust nano and capillary flow into your LC-MS using Thermo Scientific™ EASY-Spray™ HPLC Columns. The integrated column/emitter design eliminates dead volume and is temperature-controlled for maximum reliability and performance. Rigorously tested to ensure maximum quality, these columns deliver maximum simplicity and ease-of-use. The capillary flow HPLC columns provide sensitive protein, peptide, and monoclonal antibody (mAb) separation. They give proteomics researchers more than ever before: more throughput, more sensitivity, more separation power and more ease of use.
The unique EASY-Spray design provides uncompromised performance and ease-of-use in nano and capillary LC-MS proteomics applications. Features include the following:
- Precision positioned glass emitter
- Integrated precision designed union
- Integrated temperature control
Specifications
Specifications
| Column Format | Analytical Column, EASY-Spray |
| Diameter (Metric) | 75 μm |
| Length (Metric) | 150 mm |
| Max. Pressure | 7250 psi (500 bar) |
| Particle Size | 5 μm |
| pH | 2 to 8 |
| Pore Size | 300 Å |
| Surface Area | 100 m²/g |
| Temperature | 60°C |
| USP Type | L1 |
| Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.