missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Red Blood Cell Lysis Buffer

Product Code. 17138021
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
100 mL
Unit Size:
100mL
10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17138021 100 mL 100mL
17108021 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17138021 Supplier STEMCELL Technologies Supplier No. 20120

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Red blood cell lysis buffer, 10X concentrate

  • EasySep™ Red Blood Cell Lysis Buffer is for use as directed with EasySep™ or RoboSep™ Whole Blood kits for the lysis of red blood cells prior to separation from whole blood

Specifications

Description Chimerism; HLA; Immunology
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.