missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eaton's Reagent, P{2}O{5} ca 7.5% w/w

Product Code. 11368346
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 11368346

missing translation for 'mfr': Thermo Scientific Chemicals L15607.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39394-84-8 | CH4O8P2S | 238.043 g/mol

Eaton's Reagent is used in the synthesis fluorine-based hole-transporting material, mono and bis-chalcone derivatives via Claisen-Schmidt condensation reaction between arylaldehydes and ketones and 4-hydroxycoumarins and 4-hydroxy-2-quinolinones derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39394-84-8
Molecular Formula CH4O8P2S
Molecular Weight (g/mol) 238.043
MDL Number MFCD00070545
InChI Key JHNLZOVBAQWGQU-UHFFFAOYSA-N
Synonym eaton's reagent, phosphorus pentoxide methanesulfonic acid, diphosphooxidane; methanesulfonic acid, eaton's, eaton'sreagent, eaton inverted exclamation markas reagent, eaton's reagent, p2o5 ca w/w, phosphorus pentoxide, 7.7 wt. % in methanesulfonic acid
PubChem CID 16149098
SMILES CS(=O)(=O)O.O=P(=O)OP(=O)=O

Specifications

Linear Formula P2O5
UN Number UN3265
Density 1.515
Solubility Information Decomposes in water.
Formula Weight 141.95
Refractive Index 1.435
Quantity 100 mL
Flash Point >110°C (230°F)
Sensitivity Moisture sensitive; May form a precipitate
Assay ≈7.5% w/w
Chemical Name or Material Eaton's Reagent, P2O5
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.