missing translation for 'onlineSavingsMsg'
Learn More

Eaton's reagent

Product Code. 10423165 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10423165 2.5 L 2.5 L
10611473 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10423165 Supplier Thermo Scientific Acros Supplier No. 369800025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39394-84-8 | O10P4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Eaton's reagent
Color Undesignated
CAS 39394-84-8, 75-75-2, 1314-56-3
Boiling Point 122.0°C (1.0 mmHg)
Molecular Formula O10P4
MDL Number MFCD00070545
Concentration 6.7 to 8.2 %
Fieser 05,535; 07,291; 08,400; 11,428
Density 1.5000g/mL
Synonym Phosphorus pentoxide, 7.5 wt.% solution in methanesulfonic acid
Formula Weight 283.89
Percent Purity 6.7 to 8.2%
Packaging Glass bottle
Refractive Index 1.4330 to 1.4360
Specific Gravity 1.5
Quantity 2.5 L
Flash Point >110°C
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Eaton's reagent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H290-May be corrosive to metals.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.