Learn More
EB3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-84924
 View more versions of this product
Product Code. 18771724
  • 572.00 € / 0.10mL
Description
EB3 Polyclonal specifically detects EB3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EB3
Polyclonal
IgG
Western Blot
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.