missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBF-1 Antibody (SR2090), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | EBF-1 |
|---|---|
| Clone | SR2090 |
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18630734
|
Novus Biologicals
NBP3-22035-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18607514
|
Novus Biologicals
NBP3-22035-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EBF-1 Monoclonal antibody specifically detects EBF-1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| EBF-1 | |
| Western Blot 1:500-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 1879 | |
| IgG | |
| Affinity purified |
| SR2090 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| COE1, Collier, Olf and EBF transcription factor 1, Early B-cell factor, early B-cell factor 1, EBFFLJ41763, O/E-1, OE-1, OLF1FLJ39389, olfactory neuronal transcription factor 1, transcription factor COE1 | |
| A synthesized peptide derived from human EBF-1 (Uniprot #: Q9UH73) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title