missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECHDC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12637-100ul
This item is not returnable.
View return policy
Description
ECHDC1 Polyclonal antibody specifically detects ECHDC1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| ECHDC1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5uL/mg of uLysate, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| dJ351K20.2, DKFZp762M1110, enoyl CoA hydratase domain containing 1, enoyl Coenzyme A hydratase domain containing 1, enoyl-CoA hydratase domain-containing protein 1, FLJ40827 | |
| Produced in rabbits immunized with E. coli-derived Human ECHDC1 fragment | |
| 100 μg | |
| Lipid and Metabolism | |
| 55862 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur