missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ECRG4 Polyclonal Antibody, Invitrogen™
Description
Probable hormone that may attenuate cell proliferation and induce senescence of oligodendrocyte and neural precursor cells in the central nervous system. ECRG4-induced senescence is characterized by G1 arrest, RB1 dephosphorylation and accelerated CCND1 and CCND3 proteasomal degradation. [UniProt].
Specifications
Specifications
| Antigen | ECRG4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | C2orf40 |
| Gene Accession No. | Q9H1Z8 |
| Gene Alias | Augurin; C2orf40; chromosome 2 open reading frame 40; ECRG4; esophageal cancer related gene 4 protein; esophageal cancer-related gene 4 protein; UNQ761/PRO1508 |
| Gene Symbols | C2orf40 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?