missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EDA-A2 Monoclonal Antibody (174813)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA562009
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Cytokine which is involved in epithelial-mesenchymal signaling during morphogenesis of ectodermal organs. Functions as a ligand activating the DEATH-domain containing receptors EDAR and EDA2R. May also play a role in cell adhesion.
Specifications
| EDA-A2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q92838 | |
| EDA | |
| Mouse myeloma cell line NS0-derived recombinant human EDA-A2/Ectodysplasin A2, Ala179-Ser389. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 174813 | |
| Unconjugated | |
| EDA | |
| Ectodysplasin A2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1896 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction