missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EDA-A2 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5149491

Product Code. 30536192

  • 429.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

Cytokine which is involved in epithelial-mesenchymal signaling during morphogenesis of ectodermal organs. Functions as a ligand activating the DEATH-domain containing receptors EDAR and EDA2R. May also play a role in cell adhesion.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EDA-A2
Polyclonal
Unconjugated
EDA
Ectodysplasin A2
Goat
Antigen affinity chromatography
RUO
1896
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
ELISA, Inhibition Assays
0.2 mg/mL
PBS with trehalose and no preservative
Q92838
EDA
Mouse myeloma cell line NS0-derived recombinant human EDA-A2/Ectodysplasin A2, Ala179-Ser389.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.