missing translation for 'onlineSavingsMsg'
Learn More

Edaravone, MedChemExpress

Product Code. 16410778
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16410778 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16410778 Supplier Medchem Express Supplier No. HYB0099
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Edaravone is a strong novel free radical scavenger, and inhibits MMP-9-related brain hemorrhage in rats treated with tissue plasminogen activator.

Edaravone is a strong novel free radical scavenger, and inhibits MMP-9-related brain hemorrhage in rats treated with tissue plasminogen activator.

Specifications

Chemical Name or Material Edaravone
Physical Form Solid
CAS 89-25-8
Color Yellow
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C10H10N2O
For Use With (Application) Neuroscience-Neurodegeneration
Synonym MCI-186
Solubility Information DMSO : 125 mg/mL (717.57 mM; Need ultrasonic)
SMILES O=C1N(C2=CC=CC=C2)N=C(C)C1
Molecular Weight (g/mol) 174.2
Formula Weight 174.2
Percent Purity 98.6%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.