Learn More
Edetate DiSodium TS Solution According to United States Pharmacopoeia (USP), Reagecon
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Description
- United States Pharmacopoeia (USP) Reference Standard, United States Pharmacopoeia (USP) Reference Material
- Specified for use in individual United States Pharmacopoeia (USP) Monographs.
- Tested and certified by an ISO17025 Accredited Test Method (INAB Ref. 264T).
- NIST traceable.
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Specifications
| Packaging | HDPE Bottle GL45 |
| Physical Form | Liquid |
| Industry Type | Product Monographs |
| Quantity | 500 mL |