missing translation for 'onlineSavingsMsg'
Learn More

EDTA disodium salt solution, Volumetric, Reag. Ph. Eur., 0.1 M EDTA-Na2, for complexometry, Solstice

Product Code. 15604280 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
6 x 500 mL
10 L
6 x 1 L
Unit Size:
1 L
10 L
2.5 L
5 L
500mL
Pack of 6
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15604280 2.5 L 2.5 L
15614280 500 mL 500mL
15694270 1 L 1 L
15654280 6 x 500 mL Pack of 6
15635735 5 L 5 L
15624280 5 L 5 L
15644280 6 x 1 L Pack of 6
15625735 10 L 10 L
15674270 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 15604280 Supplier Honeywell-Fluka Supplier No. 345502.5L
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Volumetric, Reag. Ph. Eur., 0.1 M EDTA-Na2, for complexometry

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethylenediaminetetraacetic acid disodium salt solution
CAS 139-33-3
Molecular Formula C10H14N2Na2O8
Linear Formula [-CH2N(CH2CO2Na)CH2CO2H]2
MDL Number MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
UN Number NONH for all modes of transport
Beilstein 3822669
Synonym ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
InChI Key ZGTMUACCHSMWAC-UHFFFAOYSA-L
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
IUPAC Name disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
Molecular Weight (g/mol) 336.21
PubChem CID 57339238
ChEBI CHEBI:64734
Formula Weight 336.21g/mol
Quantity 2.5 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.